missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Bromoethyl)phthalimide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 82°C |
| Color | White-Yellow |
| Boiling Point | 318°C |
| Quantity | 25 g |
| Formula Weight | 254.08 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(2-Bromoethyl)phthalimide |
Safety and Handling
| TSCA | Yes |