missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromoethyl)phthalimide 98.0+%, TCI America™

Catalog No. B059725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059725G 25 g
B0597100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B059725G Supplier TCI America Supplier No. B059725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 574-98-1
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
MDL Number MFCD00005902
InChI Key CHZXTOCAICMPQR-UHFFFAOYSA-N
Synonym n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione
PubChem CID 11325
IUPAC Name 2-(2-bromoethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr

Specifications

Melting Point 82°C
Color White-Yellow
Boiling Point 318°C
Quantity 25 g
Formula Weight 254.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Bromoethyl)phthalimide