missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)pyrrolidine, 99%

Catalog No. AC103700050
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103700050 5 mL Glass bottle
AC103700250 25 mL Glass bottle
AC103701000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103700050 Supplier Thermo Scientific Chemicals Supplier No. 103700050

CAS: 7154-73-6 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7154-73-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00003182
InChI Key WRXNJTBODVGDRY-UHFFFAOYSA-N
Synonym 1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
PubChem CID 1344
IUPAC Name 2-pyrrolidin-1-ylethanamine
SMILES C1CCN(C1)CCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 0.9000g/mL
Boiling Point 66°C to 70°C (23.0 mmHg)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4677 to 1.4697
Quantity 5 mL
Specific Gravity 0.9
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)pyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-509-5

RUO – Research Use Only