missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Aminoethyl)piperazine 99.0+%, TCI America™

Catalog No. A030425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A030425ML 25 mL
A0304500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A030425ML Supplier TCI America Supplier No. A030425ML

Chemical Identifiers

CAS 140-31-8
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.21
MDL Number MFCD00005971
InChI Key IMUDHTPIFIBORV-UHFFFAOYSA-N
Synonym n-aminoethylpiperazine, n-2-aminoethyl piperazine, 1-2-aminoethyl piperazine, 2-piperazin-1-yl ethanamine, 1-piperazineethanamine, aminoethylpiperazine, 1-piperazineethylamine, 1-aminoethylpiperazine, piperazineethanamine, piperazine, 1-2-aminoethyl
PubChem CID 8795
IUPAC Name 2-(piperazin-1-yl)ethan-1-amine
SMILES NCCN1CCNCC1

Specifications

Melting Point -19°C
Color Yellow
Boiling Point 222°C
Quantity 25 mL
UN Number 2815
Formula Weight 129.21
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Aminoethyl)piperazine
EINECSNumber (5)-0961
RTECSNumber TK8050000
TSCA Yes