missing translation for 'onlineSavingsMsg'
Learn More

N-2-Acetylguanine, 99%

Catalog No. AC258400010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258400010 1 g Glass bottle
AC258400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC258400010 Supplier Thermo Scientific Chemicals Supplier No. 258400010

CAS: 19962-37-9 | C7H7N5O2 | 193.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19962-37-9
Molecular Formula C7H7N5O2
Molecular Weight (g/mol) 193.17
MDL Number MFCD00078201
InChI Key MXSMRDDXWJSGMC-UHFFFAOYSA-N
Synonym 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide
PubChem CID 88319
IUPAC Name N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N=CN2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >260°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in acids and alkalies
Formula Weight 193.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-2-Acetylguanine
Show More Show Less
EINECSNumber 243-443-7

RUO – Research Use Only