missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Acetamido)iminodiacetic Acid 98.0+%, TCI America™

Catalog No. A0699500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0699500G 500 g
A069925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0699500G Supplier TCI America Supplier No. A0699500G
Only null left

Chemical Identifiers

CAS 26239-55-4
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.155
MDL Number MFCD00008031
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
Synonym n-2-acetamido iminodiacetic acid, 2,2'-2-amino-2-oxoethyl azanediyl diacetic acid, carbamoylmethylaminodiacetic acid, ada, 2,2'-2-amino-2-oxoethyl imino diacetic acid, n-carbamoylmethyl iminodiacetic acid, glycine, n-2-amino-2-oxoethyl-n-carboxymethyl, n-carbamoylmethyliminodi acetic acid, unii-5c4r3o704e
PubChem CID 117765
ChEBI CHEBI:43960
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 201°C
Color White
Quantity 500 g
Formula Weight 190.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Acetamido)iminodiacetic Acid
EINECSNumber (2)-3614
TSCA Yes