missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Acetamido)-2-aminoethanesulfonic Acid 99.0+%, TCI America™

Catalog No. A070025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A070025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A070025G Supplier TCI America Supplier No. A070025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym aces, n-carbamoylmethyl taurine, n-2-acetamido-2-aminoethanesulfonic acid, 2-2-amino-2-oxoethyl amino ethanesulfonic acid, ethanesulfonic acid, 2-2-amino-2-oxoethyl amino, unii-i5spl5yvby, i5spl5yvby, 2-carbamoylmethylamino ethanesulfonic acid
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N

Specifications

Color White
Quantity 25 g
Formula Weight 182.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Acetamido)-2-aminoethanesulfonic Acid [Good′s buffer component for biological research]