missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dimethylphenyl)piperidine-2-carboxamide 98.0+%, TCI America™

Catalog No. d434825g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D434825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D434825G Supplier TCI America Supplier No. D434825G

Chemical Identifiers

CAS 15883-20-2
Molecular Formula C14H20N2O
Molecular Weight (g/mol) 232.327
MDL Number MFCD01701244
InChI Key SILRCGDPZGQJOQ-UHFFFAOYSA-N
Synonym N-Desbutyl Bupivacaine
PubChem CID 115282
IUPAC Name N-(2,6-dimethylphenyl)piperidine-2-carboxamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)C2CCCCN2

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 232.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
RTECSNumber TM6076000
TSCA No