missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Diethylphenyl)maleimide, 97%

Catalog No. aab2065606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2065606 5 g
AAB2065603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2065606 Supplier Thermo Scientific Chemicals Supplier No. B2065606
Only null left

CAS: 38167-72-5 | C14H15NO2 | 229.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38167-72-5
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.279
MDL Number MFCD00173826
InChI Key LNOKVKHZEYOLIQ-UHFFFAOYSA-N
Synonym n-2,6-diethylphenyl maleimide, 1-2,6-diethylphenyl-1h-pyrrole-2,5-dione, 1-2,6-diethylphenyl pyrrole-2,5-dione, 1-2,6-diethylphenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-2,6-diethylphenyl azoline-2,5-dione, acmc-1ctvb, cyto10h11, maybridge1_007020, 1h-pyrrole-2, 1-2,6-diethylphenyl, 1h-pyrrole-2,5-dione,1-2,6-diethylphenyl
PubChem CID 300890
IUPAC Name 1-(2,6-diethylphenyl)pyrrole-2,5-dione
SMILES CCC1=C(C(=CC=C1)CC)N2C(=O)C=CC2=O

Specifications

Quantity 5 g
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material N-(2,6-Diethylphenyl)maleimide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only