missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-phenylalanine 98.0+%, TCI America™

Catalog No. D1035100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1035100MG 100 mg
D10351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1035100MG Supplier TCI America Supplier No. D1035100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1655-54-5
Molecular Formula C15H13N3O6
Molecular Weight (g/mol) 331.28
MDL Number MFCD00038099
InChI Key HJQHTLAEPSKXQJ-UHFFFAOYNA-N
Synonym N-Dnp-L-phenylalanine, Dnp-Phe-OH
PubChem CID 7266694
IUPAC Name 2-[(2,4-dinitrophenyl)amino]-3-phenylpropanoic acid
SMILES OC(=O)C(CC1=CC=CC=C1)NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 187°C
Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 331.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-phenylalanine