missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitrophenyl)-L-leucine 99.0+%, TCI America™

Catalog No. D10331G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10331G 1 g
D1033100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10331G Supplier TCI America Supplier No. D10331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1655-57-8
Molecular Formula C12H15N3O6
Molecular Weight (g/mol) 297.267
MDL Number MFCD00038102
InChI Key STMDPCBYJCIZOD-JTQLQIEISA-N
Synonym N-Dnp-L-leucine, Dnp-Leu-OH
PubChem CID 15487857
SMILES CC(C)CC(C(=O)O)NC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 98°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 297.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-leucine