missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitro-5-fluorophenyl)-L-valinamide, For chiral derivatization, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111012947
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012947 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012947 Supplier Merck Emd Millipore Supplier No. 42102100MG

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide [L-FDVA] is a derivatizing chiral agent.

Chemical Identifiers

MDL Number MFCD00155636

Specifications

Melting Point 173°C to 177°C; 173°C to 177°C
CAS 132679-61-9
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula FC6H2(NO2)2 NH[CHCH(CH3)2 ]CONH2
Quantity 100 mg
Formula Weight 300.24
Percent Purity ≥98.0% (sum of enantiomers, HPLC); ≥98.0%
Grade For chiral derivatization