missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dinitro-5-fluorophenyl)-D-valinamide, For chiral derivatization, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111013527
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013527 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013527 Supplier Merck Emd Millipore Supplier No. 42100500MG

Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide [D-FDVA] is a derivatizing agent.

Chemical Identifiers

MDL Number MFCD00155643

Specifications

Melting Point 173°C to 177°C
CAS 210529-62-7
Linear Formula C11H13 FN4O5
Quantity 500 mg
Formula Weight 300.24
Percent Purity ≥98.0% (sum of enantiomers, HPLC); ≥98.0%
Grade For chiral derivatization