missing translation for 'onlineSavingsMsg'
Learn More

N,2,3-Trimethyl-2-isopropylbutamide

Catalog No. AC421930250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421930250 25 g Glass bottle
AC421930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421930250 Supplier Thermo Scientific Chemicals Supplier No. 421930250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51115-67-4 | C10H21NO | 171.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51115-67-4
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00130070
InChI Key RWAXQWRDVUOOGG-UHFFFAOYSA-N
Synonym 2-isopropyl-n,2,3-trimethylbutyramide, 2-isopropyl-n,2,3-trimethylbutanamide, trimethyl isopropyl butanamide, methyl diisopropyl propionamide, n,2,3-trimethyl-2-1-methylethyl butanamide, unii-6qop5a9489, n,2,3-trimethyl-2-isopropylbutanamide, n,2,3-trimethyl-2-isopropylbutamide, butanamide, n,2,3-trimethyl-2-1-methylethyl, n,2,3-trimethyl-2-propan-2-yl butanamide
PubChem CID 65300
IUPAC Name N,2,3-trimethyl-2-propan-2-ylbutanamide
SMILES CNC(=O)C(C)(C(C)C)C(C)C

Specifications

Melting Point 58.0°C to 61.0°C
Packaging Glass bottle
Quantity 25 g
Merck Index 15,9887
Formula Weight 171.28
Chemical Name or Material N, 2, 3-Trimethyl-2-isopropylbutamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 256-974-4

RUO – Research Use Only