Learn More
N-(2,3-Epoxypropyl)phthalimide, 95%
CAS: 5455-98-1 | C11H9NO3 | 203.20 g/mol
$282.38 - $282.38
Chemical Identifiers
| CAS | 5455-98-1 |
|---|---|
| Molecular Formula | C11H9NO3 |
| Molecular Weight (g/mol) | 203.20 |
| MDL Number | MFCD00005896,MFCD04973349,MFCD04973350 |
| InChI Key | DUILGEYLVHGSEE-UHFFFAOYNA-N |
| Synonym | n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl |
| PubChem CID | 21601 |
| IUPAC Name | 2-(oxiran-2-ylmethyl)isoindole-1,3-dione |
| SMILES | O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC170040100
|
Thermo Scientific Chemicals
170040100 |
10 g | Glass Bottle |
Each for $282.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5455-98-1 | |
| 203.20 | |
| DUILGEYLVHGSEE-UHFFFAOYNA-N | |
| 21601 | |
| O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 |
| C11H9NO3 | |
| MFCD00005896,MFCD04973349,MFCD04973350 | |
| n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl | |
| 2-(oxiran-2-ylmethyl)isoindole-1,3-dione |
Specifications
| 5455-98-1 | |
| 100.0 | |
| White to Yellow | |
| 94% min. (GC) | |
| C11H9NO3 | |
| 10 g | |
| DUILGEYLVHGSEE-UHFFFAOYNA-N | |
| 2-(oxiran-2-ylmethyl)isoindole-1,3-dione | |
| 21601 | |
| 95% | |
| N-(2, 3-Epoxypropyl)phthalimide |
| 94.0 | |
| 94.0°C to 100.0°C | |
| Authentic | |
| Glass Bottle | |
| MFCD00005896,MFCD04973349,MFCD04973350 | |
| n-2,3-epoxypropyl phthalimide, n-glycidylphthalimide, 2-oxiran-2-ylmethyl isoindoline-1,3-dione, 2-oxiran-2-ylmethyl-1h-isoindole-1,3 2h-dione, 2,3-epoxypropylphthalimide, 1h-isoindole-1,3 2h-dione, 2-oxiranylmethyl, denacol ex 731, phthalimidomethyl oxirane, n-2,3-epoxypropyl-2-phthalimide, phthalimide, n-2,3-epoxypropyl | |
| O=C1N(CC2CO2)C(=O)C2=CC=CC=C12 | |
| 203.20 | |
| 203.2 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes,on skin,or on clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
EINECSNumber : 226-710-2
RUO – Research Use Only