missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1-Naphthyl)formamide 98.0+%, TCI America™
Supplier: TCI America N100125G
Specifications
| N-(1-Naphthyl)formamide | |
| 140°C | |
| MFCD00086455 | |
| CGRYTQQVSFZYCI-UHFFFAOYSA-N | |
| N-(naphthalen-1-yl)formamide | |
| 96004 | |
| ≥98.0% (GC) |
| 6330-51-4 | |
| C11H9NO | |
| 25 g | |
| O=CNC1=C2C=CC=CC2=CC=C1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
Chemical Identifiers
| 6330-51-4 | |
| 171.20 | |
| CGRYTQQVSFZYCI-UHFFFAOYSA-N | |
| N-(naphthalen-1-yl)formamide |
| C11H9NO | |
| MFCD00086455 | |
| 96004 | |
| O=CNC1=C2C=CC=CC2=CC=C1 |
Safety and Handling
TSCA : No