missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride monoMethanolate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013003
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013003 25 g
111013004 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013003 Supplier Merck Emd Millipore Supplier No. 7072025G

N-(1-Naphthyl)ethylenediamine dihydrochloride is a hygroscopic analytical reagent generally suitable for determination of drugs, pharmaceuticals with free primary aromatic amino group, sulpha drugs and local anaesthetics. It may decompose on exposure to light.

Chemical Identifiers

MDL Number MFCD16661167

Specifications

Melting Point 196°C to 199°C
CAS 1357471-44-3
Linear Formula C10H7 NHCH2 CH2 NH2 · 2 HCl · MeOH
Quantity 25 g
Formula Weight 291.22
Percent Purity ≥99.0% (AT); ≥99.0%
Grade For Spectrophotometric Det. of Nitrate and Nitrite