missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride for determination of sulfonamide and nitrite, ACS reagent, ≥98%, MilliporeSigma™ Supelco™

Catalog No. p-7187633
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013140 5 g
111013139 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013140 Supplier Merck Emd Millipore Supplier No. 334615G
Only null left

Chemical Identifiers

MDL Number MFCD00012556
Synonym 2-(1-Naphthylamino)ethylamine dihydrochloride

Specifications

Melting Point 194°C to 198°C
CAS 1465-25-4
Linear Formula C10H7 NHCH2 CH2 NH2 · 2 HCl
Quantity 5 g
Formula Weight 259.17
Percent Purity ≥98% (calc with ref. to anhyd. subst.); ≥98%
Grade ACS Reagent
RTECSNumber KV5330000