missing translation for 'onlineSavingsMsg'
Learn More

N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7127670
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D333125G 25 g
D3331500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D333125G Supplier TCI America Supplier No. D333125G

Chemical Identifiers

CAS 793-24-8
Molecular Formula C18H24N2
Molecular Weight (g/mol) 268.404
InChI Key ZZMVLMVFYMGSMY-UHFFFAOYSA-N
PubChem CID 13101
IUPAC Name 4-N-(4-methylpentan-2-yl)-1-N-phenylbenzene-1,4-diamine
SMILES CC(C)CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 51°C
Boiling Point 165°C
Quantity 25 g
UN Number 3077
Synonym 4-[(4-Methyl-2-pentyl)amino]diphenylamine, N-(4-Methyl-2-pentyl)-N′C-phenyl-1,4-phenylenediamine
Formula Weight 268.40
Percent Purity ≥98.0% (GC)
Physical Form Pellets
Chemical Name or Material N-(1,3-Dimethylbutyl)-N′-phenyl-1,4-phenylenediamine
EINECSNumber (3)-0136&(3)-0368
RTECSNumber ST0900000
TSCA Yes