missing translation for 'onlineSavingsMsg'
Learn More

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 15205-27-3
Molecular Formula C9H12NO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD04496422
InChI Key CEPGPPSMCRKGFJ-UHFFFAOYSA-O
Synonym n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
PubChem CID 421238
IUPAC Name 1-(1,3-benzodioxol-5-yl)-N-methylmethanamine
SMILES C[NH2+]CC1=CC=C2OCOC2=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC01646CB
SDS View Documents
Thermo Scientific Chemicals
CC01646CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01646DA
SDS View Documents
Thermo Scientific Chemicals
CC01646DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC01646EA
SDS View Documents
Thermo Scientific Chemicals
CC01646EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

15205-27-3
166.20
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
421238
C[NH2+]CC1=CC=C2OCOC2=C1
C9H12NO2
MFCD04496422
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
1-(1,3-benzodioxol-5-yl)-N-methylmethanamine

Specifications

15205-27-3
C9H12NO2
250mg
CEPGPPSMCRKGFJ-UHFFFAOYSA-O
[(2H-1,3-benzodioxol-5-yl)methyl](methyl)azanium
421238
97%
97%
MFCD04496422
n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
C[NH2+]CC1=CC=C2OCOC2=C1
166.20
165.19
N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
Videos
SDS
Documents

Documents