missing translation for 'onlineSavingsMsg'
Learn More

Myristic Acid 99.0+%, TCI America™

Catalog No. M047625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M047625G 25 g
M0476500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M047625G Supplier TCI America Supplier No. M047625G

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 54°C
Color White
Boiling Point 326°C
Quantity 25 g
Formula Weight 228.38
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Myristic Acid
EINECSNumber (2)-0608
RTECSNumber QH4375000
TSCA Yes