missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Myristic acid, 99%

Catalog No. p-3593468
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156961000 100 g Glass bottle
AC156962500 250 g Glass bottle
AC156960010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156961000 Supplier Thermo Scientific Chemicals Supplier No. 156961000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 544-63-8
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00002744
InChI Key TUNFSRHWOTWDNC-UHFFFAOYSA-N
Synonym myristic acid, n-tetradecanoic acid, n-tetradecoic acid, crodacid, n-tetradecan-1-oic acid, 1-tridecanecarboxylic acid, hydrofol acid 1495, univol u 316s, emery 655, myristinsaeure
PubChem CID 11005
ChEBI CHEBI:28875
IUPAC Name tetradecanoic acid
SMILES CCCCCCCCCCCCCC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 57.0°C
Color White
Boiling Point 326.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)12CO2H
Quantity 100 g
Beilstein 02, 365
Merck Index 15, 6419
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, methanol, ether, petr.ether,, benzene and chloroform
Formula Weight 228.37
Percent Purity 99%
Physical Form Crystalline Mass, Powder, Flakes or Chunks
Chemical Name or Material Myristic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len
EINECSNumber 208-875-2

RUO – Research Use Only