missing translation for 'onlineSavingsMsg'
Learn More

Myrcene (stabilized with BHT) 75.0+%, TCI America™

Catalog No. M023525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M023525ML 25 mL
M0235500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M023525ML Supplier TCI America Supplier No. M023525ML

Chemical Identifiers

CAS 123-35-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00008908
InChI Key UAHWPYUMFXYFJY-UHFFFAOYSA-N
Synonym myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
PubChem CID 31253
ChEBI CHEBI:17221
IUPAC Name 7-methyl-3-methylideneocta-1,6-diene
SMILES CC(=CCCC(=C)C=C)C

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 3295
Formula Weight 136.24
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material Myrcene (stabilized with BHT)
EINECSNumber (2)-2358
RTECSNumber RG5365000
TSCA Yes
Recommended Storage Refrigerator