missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MyPIPETMANTM Volume Display Windows

Catalog No. FP070104G
Change view
Click to view available options
Color:
Blue
Green
Navy
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
FP070104G Red
FP070105G Yellow
FP070106G Green
FP070103G Blue
FP070101G Navy
FP070102G White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. FP070104G Supplier Gilson™ Supplier No. FP070104

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For easier identification and convenience, this colored window provides extra pipette customization.

  • Six available colors
  • Easily replace the colored window at your convenience

Specifications

Autoclavable Autoclavable
Calibrated No
Material Polystyrene, Polypropylene
Sterility Non-sterile
Graduated No
Quantity 1
Color Red
Type Display Window
Disposable Reusable
For Use With (Equipment) MyPIPETMAN