missing translation for 'onlineSavingsMsg'
Learn More

Myosmine, 98%, may contain up to 2% water

Catalog No. AAL11886ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11886ME 500 mg
AAL11886MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL11886ME Supplier Thermo Scientific Chemicals Supplier No. L11886ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 532-12-7 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532-12-7
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00052019
InChI Key DPNGWXJMIILTBS-UHFFFAOYSA-N
Synonym myosmine, 3-3,4-dihydro-2h-pyrrol-5-yl pyridine, 3-1-pyrrolin-2-yl pyridine, miosmine, unii-9o0a545w4l, 3-4,5-dihydro-3h-pyrrol-2-yl-pyridine, 2-3-pyridyl-1-pyrroline, 3-2-pyrrolin-2-yl pyridine, pyridine, 3-3,4-dihydro-2h-pyrrol-5-yl, pyridine, 3-1-pyrrolin-2-yl
PubChem CID 442649
ChEBI CHEBI:7051
IUPAC Name 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
SMILES C1CC(=NC1)C2=CN=CC=C2

Specifications

Melting Point 42°C to 44°C
Boiling Point 82°C to 83°C (0.5 mmHg)
Quantity 500 mg
Beilstein 119254
Formula Weight 146.19
Percent Purity 98%
Assay may contain up to 2% water
Chemical Name or Material Myosmine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
RTECSNumber UT7660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only