missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ MYH Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 4595 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB600-1032 |
| Gene Alias | EC 3.2.2, EC 3.2.2.-, EC 3.2.2.31, hMYH, MGC4416, MutY homolog, MUTYH, Muyth, MYH, MYHA/G-specific adenine DNA glycosylase |
| Gene Symbol | MUTYH |
| Quantity | 0.1 mg |
| Research Category | Base Excision Repair, Cancer, DNA Repair |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |