missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycophenolic Acid, MP Biomedicals
$515.51 - $515.51
Chemical Identifiers
| CAS | 24280-93-1 |
|---|---|
| Molecular Formula | C17H20O6 |
| Molecular Weight (g/mol) | 320.341 |
| MDL Number | MFCD00036814 |
| InChI Key | HPNSFSBZBAHARI-RUDMXATFSA-N |
| Synonym | mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid |
| PubChem CID | 446541 |
| ChEBI | CHEBI:168396 |
| IUPAC Name | (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid |
| SMILES | CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19417283
|
MP Biomedicals Inc
0219417283 |
250 mg |
Each for $515.51
|
|
|||||
Chemical Identifiers
| 24280-93-1 | |
| 320.341 | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| 446541 | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid |
| C17H20O6 | |
| MFCD00036814 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| CHEBI:168396 | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC |
Specifications
| 24280-93-1 | |
| ≥98% | |
| MFCD00036814 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC | |
| 320.341 | |
| CHEBI:168396 | |
| Powder |
| White | |
| C17H20O6 | |
| 250 mg | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid | |
| 446541 | |
| 320.3 | |
| Mycophenolic Acid, From Penicillium brevi compactum |
Safety and Handling
EINECSNumber : 246-119-3