Learn More
Mycophenolic acid, 98%
CAS: 24280-93-1 | C17H20O6 | 320.341 g/mol
Supplier: Thermo Scientific Chemicals J6190509
Description
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.
Solubility
Soluble in methanol at 50mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agent.
Specifications
| Mycophenolic acid | |
| 139°C to 141°C | |
| C17H20O6 | |
| 10 g | |
| 14,6327 | |
| Soluble in methanol at 50mg/ml | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC | |
| 320.341 | |
| CHEBI:168396 | |
| 98% |
| 24280-93-1 | |
| White | |
| MFCD00036814 | |
| 1295848 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid | |
| 446541 | |
| 320.34 | |
| Powder |
Chemical Identifiers
| 24280-93-1 | |
| 320.341 | |
| HPNSFSBZBAHARI-RUDMXATFSA-N | |
| 446541 | |
| (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid |
| C17H20O6 | |
| MFCD00036814 | |
| mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid | |
| CHEBI:168396 | |
| CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H360D-H372
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 246-119-3
RTECSNumber : MP8050000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only