missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mycophenolic acid, 98%

Catalog No. AC459380250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459380250 25 g
AC459380010 1 g
AC459380050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459380250 Supplier Thermo Scientific Chemicals Supplier No. 459380250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24280-93-1 | C17H20O6 | 320.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24280-93-1
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.34
InChI Key HPNSFSBZBAHARI-RUDMXATFSA-N
Synonym mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid
PubChem CID 446541
ChEBI CHEBI:168396
IUPAC Name (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 320.34
Percent Purity 98%
Chemical Name or Material Mycophenolic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May damage the unborn child.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long last
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Use personal protective equipment as required.
IF exposed or concerned: Get m
EINECSNumber 246-119-3

RUO – Research Use Only