Learn More
Innovatek MycoPak™ Dermatophyte Specimen Collection Kit
Catalog No. 361041863
Change view
Click to view available options
Quantity:
100 Kits/Box
Catalog No. Quantity
361041863 100 Kits/Box
1 options
Catalog No. 361041863 Supplier Innovatek Supplier No. 4001MPA100
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovatek MycoPak™ Dermatophyte Specimen Collection Kit for superficial mycology specimen samples (skin, hair, nails)
  • Collection and transport of superficial mycology specimens (skin, hair, nails)
  • Fits in a regular envelope for mailing to the lab
  • Easy and convenient for physicians to collect fungal samples, with detailed instructions
  • Multiple folds in the black specimen paper prevents loss of specimen fragments
  • Designed to retain the sample even when opened in a laboratory safety laminar flow cabinet
  • Includes specimen paper, paper envelope, plastic zip-top bag, collection instructions
Specifications
Quantity 100 Kits/Box