missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Myco-Sniff-Valid™ Mycoplasma PCR Detection Kit

Catalog No. MP093050301
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP093050301 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP093050301 Supplier MP Biomedicals™ Supplier No. 093050301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Validated for sensitive, specific and rapid detection of mycoplasma.

  • Highly sensitive: The detection limit is as low as 10 CFU/mL
  • Reliable: Meets European Pharmacopoeia Guidance and Regulation. This test is suitable for product release testing and in-process control. It can replace culture and indicator cell tests
  • Fast: Replaces traditional culture testing with 3 hours PCR test
  • Wide detection range: Detects common cell culture-infecting species of mycoplasma and also other various species of mycoplasma
  • Easy to use: Contains premixed components for the PCR reaction. Just add template DNA or samples
  • Highly specific: No interference of animal or bacterial DNA
  • Exogenous internal control: Helps differentiate false negatives due to PCR inhibition or erroneous PCR testes
  • Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA

Specifications

Content And Storage Store at -20°C
Antibiotic Spectrum Mycoplasma
Product Type Mycoplasma Detection
For Use With (Application) Mycoplasma detection
Quantity 48 Tests