missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Myco-Sniff-Rapid™ Mycoplasma Luciferase Detection Kit
Description
- A quick, simple, and sensitive method for detecting mycoplasma contamination in cell cultures
- Kit utilizes the activity of mycoplasma metabolic enzymes from 209 mycoplasma species, including those commonly found contaminating cell cultures
- Since this enzyme is not present in eukaryotic cells, the test is specific for mycoplasma
- In the presence of mycoplasma, the enzymes react with the substrate to catalyze the conversion of ADP to ATP
- Mycoplasma contamination can be detected by measuring the level of ATP in a sample before and after the addition of Myco-Sniff-Rapid™ substrate via a luciferase assay
Specifications
Specifications
| Content And Storage | Myco-Sniff-Rapid Reagent (Lyophilized), Myco-Sniff-Rapid Substrate (Lyophilized), and MycoFree Water |
| Description | Mycoplasma Detection Kit |
| Detection Method | Luciferase |
| Form | Lyophilized |
| Sterility | Non-sterile |
| For Use With | A Luminometer is Needed for Reading The Luminescent Signal. The Product is Suitable for Use with Any Tube Luminometer, Plate Luminometer, or Multifunctional Reader. |
| For Use With (Application) | Mycoplasma detection |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
For research or further manufacturing use only, not for direct human use.