missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Myco-Sniff-Rapid™ Mycoplasma Luciferase Detection Kit

Catalog No. MP093050402
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP093050402 50 Tests
MP093050401 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP093050402 Supplier MP Biomedicals™ Supplier No. 093050402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A rapid, 20 minute method that is simple and sensitive for the detection of mycoplasma contamination in cell cultures

  • A quick, simple, and sensitive method for detecting mycoplasma contamination in cell cultures
  • Kit utilizes the activity of mycoplasma metabolic enzymes from 209 mycoplasma species, including those commonly found contaminating cell cultures
  • Since this enzyme is not present in eukaryotic cells, the test is specific for mycoplasma
  • In the presence of mycoplasma, the enzymes react with the substrate to catalyze the conversion of ADP to ATP
  • Mycoplasma contamination can be detected by measuring the level of ATP in a sample before and after the addition of Myco-Sniff-Rapid™ substrate via a luciferase assay

Specifications

Content And Storage Myco-Sniff-Rapid Reagent (Lyophilized), Myco-Sniff-Rapid Substrate (Lyophilized), and MycoFree Water
Description Mycoplasma Detection Kit
Detection Method Luciferase
Form Lyophilized
Sterility Non-sterile
For Use With A Luminometer is Needed for Reading The Luminescent Signal. The Product is Suitable for Use with Any Tube Luminometer, Plate Luminometer, or Multifunctional Reader.
For Use With (Application) Mycoplasma detection
Quantity 50 Tests
Regulatory Status RUO

For research or further manufacturing use only, not for direct human use.