missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Myco-Sniff™ Mycoplasma PCR Detection Kit

Catalog No. MP093050201
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP093050201 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP093050201 Supplier MP Biomedicals™ Supplier No. 093050201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for sensitive, specific, and rapid detection of mycoplasma.

  • Highly sensitive: The detection limit is as low as 20 CFU/mL
  • Wide detection range of mycoplasma: Detect mycoplasma from over 8 genera, including 209 individual species
  • Premixed for ease-of-use: The kit contains all the components for the PCR reaction. Just add template DNA or samples
  • Highly specific: No interference of animal or bacterial DNA
  • Fast: Detection can be achieved within 3 hours
  • Elimination of cross-contamination: 8-MOP prevents cross-contamination from previous PCR products
  • Exogenous internal control: Helps differentiate false negatives due to PCR inhibition or erroneous PCR tests
  • Sample control: Easily verify the effectiveness of template gDNA by checking the amplification of the sample control DNA

Specifications

Content And Storage Store at -20°C
Antibiotic Spectrum Mycoplasma
Product Type Mycoplasma Detection
For Use With (Application) Mycoplasma detection
Quantity 48 Tests