missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MYBL1 Split FISH Probe

Catalog No. 89999064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89999064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89999064 Supplier Abnova™ Supplier No. FS0067

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

  • The MYBL1 Split FISH Probe is designed for the identification of gene splits using the Fluorescent In Situ Hybridization (FISH) technique.
  • Applications: Fluorescent In Situ Hybridization (Cell)
  • Reactivity: Human
  • Source: Genomic DNA
  • Probe 1: MYBL1, approximately 260 kb, labeled with Texas Red at 8q13.1
  • Probe 2: MYBL1, approximately 460 kb, labeled with FITC at 8q13.1
  • Supplied Products: DAPI Counterstain (1500 ng/mL), 125 μL per 100 μL of FISH probe
  • Form: Liquid
  • Storage: 4°C in the dark
  • Regulation Status: For research use only (RUO)

Specifications

Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Detection Method Fluorescence
Format Tube
Label or Dye FITC
For Use With (Application) FISH-Ce
Gene ID (Entrez) 4603
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Product Type FISH Probe
Quantity 100 μL
Target Human
Form Liquid
Show More Show Less