missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MYB/NFIB DY Translocation FISH Probe
Description
- The MYB/NFIB Probe Set is designed for the detection of the MYB/NFIB translocation and consists of DNA labeled in Spectrum Orange and Spectrum Green.
- Probe Specificity: Hybridizes to chromosome 6q23.3 (Orange) and 9p23 (Green) in normal metaphase spreads and interphase nuclei.
- Genome Build: GRCh37/hg19.
- Research Use Only: This product is for research use only and has not been validated or tested on clinical samples.
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark. |
| Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Detection Method | Fluorescence |
| Format | Tube |
| Label or Dye | FITC |
| For Use With (Application) | FISH-Ce |
| Gene ID (Entrez) | 4602/4781 |
| Includes | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
| Product Type | FISH Probe |
| Quantity | 100 μL |
| Show More |