Learn More
Epredia™ MX35 Premier™ Disposable Low-profile Microtome Blades
Catalog No. 3051835
Change view
Click to view available options
Product Type:
Microtome Blade, Premier
Microtome Blade, Premier Plus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
3051835 Microtome Blade, Premier
3052835 Microtome Blade, Premier Plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3051835 Supplier Epredia™ Supplier No. 3051835
Section hard tissues and high-quality thin sections with high-performance with Epredia™ MX35 Premier™ disposable low-profile microtome blades.
  • Sharper edge with increased durability for longer blade life
  • Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life
Specifications
Product Type Microtome Blade, Premier
Profile Low
For Use With (Application) Designed for routine histology and are good for a variety of tissue types, including hard tissue
Disposable Single-use
Cutting Angle 34°
Thickness (English) 0.01 in.
Thickness (Metric) 0.24 mm