missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Wire Shelves

Catalog No. 91138
Change view
Click to view available options
Dimensions (L x W x H):
24 x 18 x 38 in.
30 x 18 x 38 in.
36 x 18 x 38 in.
36 x 21 x 39 in.
36 x 24 x 39 in.
Handle Material:
Chrome
Stainless Steel
Shelf Material:
Chrome
Stainless Steel
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Handle Material Shelf Material
91138 30 x 18 x 38 in. Chrome Chrome
19138778 36 x 24 x 39 in. Stainless Steel Stainless Steel
19138773 36 x 18 x 38 in. Chrome Chrome
19138770 24 x 18 x 38 in. Chrome Chrome
19138776 36 x 21 x 39 in. Stainless Steel Stainless Steel
19138772 30 x 18 x 38 in. Stainless Steel Stainless Steel
19138777 36 x 24 x 39 in. Chrome Chrome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 91138 Supplier Metro™ Supplier No. MW703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.

  • Cart load capacity 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
  • One-piece stainless-steel handles add rigidity and durability
  • 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
  • Three tiers
  • Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Product Type Utility Cart
Capacity (Metric) 170 kg
Certifications/Compliance NSF
Color Silver
Description 3-tier Model
Dimensions (L x W x H) 30 x 18 x 38 in.
For Use With (Application) Transporting Materials
Handle Material Chrome
Height (English) 38 in.
Height (Metric) 97 cm
Includes Super Erecta™ 3 Chrome-plated wire shelves with plastic split sleeves, 2 one-piece Chrome-plated handle, 4 in. dia. caster, 3.5 in. donut bumpers
Length (English) 30 in.
Length (Metric) 76 cm
Load Bearing Capacity (English) 375 lb.
Load Bearing Capacity (Metric) 170 kg
Material Steel
No. of Shelves 3
Shelf Material Chrome
Width (English) 18 in.
Width (Metric) 46 cm
Show More Show Less