missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Wire Shelves

Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.

Supplier:  Metro™ MW712

 View more versions of this product

Catalog No. 19138778


Only null left

Description

Description

  • Cart load capacity 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
  • One-piece stainless-steel handles add rigidity and durability
  • 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
  • Three tiers
  • Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Chemical Identifiers

Utility Cart
NSF
3-tier Model
375 lb.
3
170 kg
Silver
Transporting Materials
170 kg

Specifications

Utility Cart
NSF
3-tier Model
Transporting Materials
39 in.
Super Erecta™ 3 Stainless Steel wire shelves with plastic split sleeves, 2 one-piece Stainless Steel handle, 5 in. dia. caster, 3.5 in. donut bumpers
91 cm
170 kg
3
24 in.
170 kg
Silver
36 x 24 x 39 in.
Stainless Steel
99 cm
36 in.
375 lb.
Stainless Steel
Stainless Steel
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions