missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Dioxide, Powder, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 2.5 kg |
| Molecular Weight (g/mol) | 86.94 |
| Grade | Technical |
| Packaging | Poly Pail |
| Chemical Name or Material | Manganese Dioxide, Powder |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.