missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ mutaFISH™ RNA Accessory Kit
mutaFISH™ RNA Accessory Kit provides reagents and enzymes for in situ reverse transcription, RNA digestion, mutaFISH™ hybridization, ligation and amplication prior to mutaFISH™.
Supplier: Abnova™ KA4915
Description
Includes:
Box 1
- DEPC-H2O
- DEPC-PBS
- PBS-T
- Permeabilization Buffer
- Formamide
- Detection Buffer
Box 2
- 3% Paraformaldehyde
- dNTP Mix
- BSA
- RNase Inhibitor
- RT Enzyme
- 5X RT Buffer
- Hybrid Enzyme
- 10X Hybrid Enzyme Buffer
- RNase H
- 1M KCL
- DNA Polymerase
- 10X DNA Polymerase Buffer
- 50% Glycerol
- DAPI (100μM)
Spécifications
| Store Box 1 at 4°C, store Box 2 at -20°C. | |
| 1 Kit |