missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ mutaFISH™ HER2wt DNA Probes

Catalog No. 89333040
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89333040 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89333040 Supplier Abnova™ Supplier No. FP0010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.

mutaFISH™ HER2wt DNA Probes is designed to identify human HER2 gene amplification on dsDNA in cells using padlock probe andin siturolling-circle amplification technology.

  • mutaFISH™ HER2wt DNA Probe 1
  • mutaFISH™ HER2wt DNA Probe 2
  • Detection Probe-Cy5

Specifications

Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Detection Method Fluorescence
Format Kit
Label or Dye Cy5
For Use With (Application) mutaFISH-Ce
Probe HER2wt
Product Type Fluorescence In Situ Hybridization
Quantity 1 Probe Set