missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ mutaFISH™ EGFR L858R L858wt Ex19wt RNA Probes

Catalog No. 89333032
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89333032 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89333032 Supplier Abnova™ Supplier No. FP0002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.

mutaFISH™ EGFR L858R L858wt Ex19wt RNA Probes is designed to detect human EGFR L858R gene mutation and EGFR exon 19 deletion on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.

Includes:

  • RT EGFR L858 Primer
  • RT EGFR Ex19 Primer
  • mutaFISH™ EGFR L858R RNA Probe
  • mutaFISH™ EGFR L858wt RNA Probe
  • mutaFISH™ EGFR Ex19wt RNA Probe
  • Detection Probe-Cy3
  • Detection Probe-Cy5
  • Detection Probe-Texas Red

Specifications

Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Label or Dye Cy3, Cy5, Texas Red
For Use With (Application) mutaFISH-Ce,mutaFISH-Fr
Location L858R,L858wt,Ex19wt
Product Type mutaFISH™ (mutation-specific Fluorescence In Situ Hybridization)
Quantity 1 Probe Set