missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ mutaFISH™ CEP7wt DNA Probes

Catalog No. 89333036
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89333036 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89333036 Supplier Abnova™ Supplier No. FP0006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.

mutaFISH™ CEP7wt DNA Probes is designed to identify human CEP7 amplification on dsDNA in cells using padlock probe and in siturolling-circle amplification technology.

Includes:

  • mutaFISH™ CEP7wt DNA Probe
  • Detection Probe-Texas Red

Specifications

Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Label or Dye Texas Red
For Use With (Application) mutaFISH-Ce
Product Type mutaFISH™ (mutation-specific Fluorescence In Situ Hybridization)
Quantity 1 Probe Set