missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ mutaFISH™ ACTBwt RNA Probes

Catalog No. 89333456
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89333456 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89333456 Supplier Abnova™ Supplier No. FP0013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

mutaFISH™ ACTBwt RNA Probes is designed to detect human ACTB wildtype gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.

Specifications

Content And Storage Store at -20°C.
Description actin, beta
Label or Dye FITC
For Use With (Application) mutaFISH-Ce
Location 7p22.1
Product Type DNA/RNA
Quantity 1 Probe Set
Size 1 Probe Set