missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscone 97.0+%, TCI America™
$262.83 - $909.67
Chemical Identifiers
| CAS | 541-91-3 |
|---|---|
| Molecular Formula | C16H30O |
| Molecular Weight (g/mol) | 238.42 |
| MDL Number | MFCD00211114 |
| InChI Key | ALHUZKCOMYUFRB-UHFFFAOYNA-N |
| Synonym | muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone |
| PubChem CID | 10947 |
| IUPAC Name | 3-methylcyclopentadecan-1-one |
| SMILES | CC1CCCCCCCCCCCCC(=O)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2611200MG
|
TCI America
M2611200MG |
200 mg |
Each for $262.83
|
|
|||||
|
M26111G
|
TCI America
M26111G |
1 g |
Each for $909.67
|
|
|||||
Chemical Identifiers
| 541-91-3 | |
| 238.42 | |
| ALHUZKCOMYUFRB-UHFFFAOYNA-N | |
| 10947 | |
| CC1CCCCCCCCCCCCC(=O)C1 |
| C16H30O | |
| MFCD00211114 | |
| muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone | |
| 3-methylcyclopentadecan-1-one |
Specifications
| 541-91-3 | |
| Colorless | |
| C16H30O | |
| 200 mg | |
| ALHUZKCOMYUFRB-UHFFFAOYNA-N | |
| 3-methylcyclopentadecan-1-one | |
| 10947 | |
| ≥97.0% (GC) | |
| Muscone |
| -15°C | |
| 95°C | |
| MFCD00211114 | |
| muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone | |
| CC1CCCCCCCCCCCCC(=O)C1 | |
| 238.42 | |
| 238.42 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-4630
RTECSNumber : GY0950000
TSCA : Yes