missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide Indicator, 0.2% (w/w) in Sodium Chloride, Ricca Chemical
Dry mixture in Sodium Chloride, APHA/EPA for Calcium Testing
Supplier: Ricca Chemical Company 5220500
Chemical Identifiers
| White Poly Wide Mouth Bottle | |
| APHA/EPA for Calcium | |
| Murexide Indicator |
Specifications
| 500 g | |
| 500 g | |
| Murexide Indicator |
| White Poly Wide Mouth Bottle | |
| APHA/EPA for Calcium |