missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Murexide Indicator, 0.15% (w/v) in Ethylene Glycol, Ricca Chemical
APHA for Calcium Testing
Supplier: Ricca Chemical Company 52214
Specifications
| 120 mL | |
| MFCD00003477 | |
| Amber Glass Bottle | |
| sodium chloride | |
| 58.44 | |
| Murexide Indicator |
| ClNa | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| [Na+].[Cl-] | |
| 120 mL | |
| APHA for Calcium |