missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Murashige and Skoog Media

Catalog No. ICN2623220
Change view
Click to view available options
Quantity:
10 x 1 L
1 x 10 L
1 x 50 L
Product Type:
Murashige and Skoog Basal Medium
Murashige and Skoog Medium
Content And Storage:
0°C to 5°C
4°C
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Content And Storage
ICN2623220 10 x 1 L Murashige and Skoog Basal Medium 0°C to 5°C
ICN2623122 1 x 10 L Murashige and Skoog Basal Medium 4°C
ICN2610020 10 x 1 L Murashige and Skoog Medium 4°C
ICN2610022 1 x 10 L Murashige and Skoog Medium 4°C
ICN2610024 1 x 50 L Murashige and Skoog Medium 4°C
ICN2623120 10 x 1 L Murashige and Skoog Basal Medium 4°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN2623220 Supplier MP Biomedicals™ Supplier No. 092623220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.

  • Each formulation is prepared according to the original specification, except where noted
  • Finished lots of media are plant cell tested

Specifications

Form Powder
Product Type Murashige and Skoog Basal Medium
With Additives Gamborg's Vitamins
Quantity 10 x 1 L
Format Solution
pH 3.75 to 4.75
Content And Storage 0°C to 5°C