Learn More
Bioworld Murashige and Skoog(MS) Stock Solution (10X), Micronutrient Only, 500 mL
Catalog No. 50254820
Encompass
Murashige & Skoog (MS) Stock Solution (10X)Contains only the Micronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 2.5–3.5Miscible in water
Catalog No. 50254820 Supplier Bioworld Supplier No. 306301281
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Murashige & Skoog (MS) Stock Solution (10X)Contains only the Micronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 2.5–3.5Miscible in water